LIF945291277 Life Chemicals Inc.

(1S,2S)-2-(methylamino)cyclopentan-1-ol

  • MDL number MFCD28151483
  • Cas Number : 934415-73-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart