LIF945291285 Life Chemicals Inc.

3-cyano-4-methylbenzoic acid

  • MDL number MFCD00045859
  • Cas Number : 138642-93-0
  • Molecular Weight : 161.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart