LIF945291288 Life Chemicals Inc.

2-cyclopropyl-2-methylpropan-1-ol

  • MDL number MFCD26000810
  • Cas Number : 1555222-37-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart