LIF945291296 Life Chemicals Inc.

1-{[(tert-butoxy)carbonyl]amino}-3-phenylcyclobutane-1-carboxylic acid

  • MDL number MFCD28389380
  • Cas Number : 1993314-86-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart