LIF945291298 Life Chemicals Inc.

4-{[(tert-butoxy)carbonyl]amino}-3-phenylbutanoic acid

  • MDL number MFCD02258141
  • Cas Number : 189014-01-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart