LIF945291310 Life Chemicals Inc.

2-methyl-1-[(2-methylpropyl)amino]propan-2-ol

  • MDL number MFCD11934337
  • Cas Number : 138001-48-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart