LIF945291319 Life Chemicals Inc.

(2R,6R)-2,6-dimethylpiperidine hydrochloride

  • MDL number MFCD27978649
  • Cas Number : 130291-36-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart