LIF945291321 Life Chemicals Inc.

pyrazolo[1,5-a]pyrazine-3-carboxylic acid

  • MDL number MFCD11865260
  • Cas Number : 53902-76-4
  • Molecular Weight : 163.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart