LIF945291342 Life Chemicals Inc.

3-cyclopropyl-1,2-oxazole-4-carboxylic acid

  • MDL number MFCD18820494
  • Cas Number : 639523-17-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart