LIF945291351 Life Chemicals Inc.

ethyl 4-[(2-hydroxyethyl)amino]piperidine-1-carboxylate

  • MDL number MFCD11154428
  • Cas Number : 1021246-92-3
  • Molecular Weight : 216.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart