LIF945291356 Life Chemicals Inc.

1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-amine hydrochloride

  • MDL number MFCD15142898
  • Cas Number : 1197238-91-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart