LIF945291358 Life Chemicals Inc.

2H,3H,5H,7H,8H-pyrano[4,3-c]pyridazin-3-one

  • MDL number MFCD16871374
  • Cas Number : 1075260-60-4
  • Molecular Weight : 152.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart