LIF945291359 Life Chemicals Inc.

5-aminopiperidin-2-one hydrochloride

  • MDL number MFCD09743647
  • Cas Number : 1235440-18-2
  • Molecular Weight : 150.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart