LIF945291365 Life Chemicals Inc.

3-(2-Phenyl-thiazol-4-yl)-phenylamine

  • MDL number MFCD02050584
  • Cas Number : 134812-32-1
  • Molecular Weight : 252.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart