LIF945291366 Life Chemicals Inc.

1-o-Tolyl-pyrimidine-2,4,6-trione

  • MDL number MFCD00552519
  • Cas Number : 107147-53-5
  • Molecular Weight : 218.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart