LIF945291371 Life Chemicals Inc.

(2-mercapto-4-methyl-6-oxo-1,6-dihydropyrimidin-5-yl)acetic acid

  • MDL number MFCD01471207
  • Cas Number : 62214-19-1
  • Molecular Weight : 200.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart