LIF945291381 Life Chemicals Inc.

2-oxocyclohexane-1-sulfonamide

  • MDL number MFCD24691452
  • Cas Number : 96355-25-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart