LIF945291472 Life Chemicals Inc.

4-ethoxybenzenesulfonamide

  • MDL number MFCD01911893
  • Cas Number : 1132-19-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart