LIF945291473 Life Chemicals Inc.

3,4-dihydro-1H-quinoxalin-2-one

  • MDL number MFCD00204133
  • Cas Number : 59564-59-9
  • Molecular Weight : 148.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart