LIF945291475 Life Chemicals Inc.

methyl 2-(morpholin-2-yl)acetate hydrochloride

  • MDL number MFCD11506162
  • Cas Number : 1187932-65-5
  • Molecular Weight : 195.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart