LIF945291476 Life Chemicals Inc.

2-hydroxypyridine-4-carbaldehyde

  • MDL number MFCD03788455
  • Cas Number : 188554-13-4
  • Molecular Weight : 123.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart