LIF945291487 Life Chemicals Inc.

(4-chlorophenyl)difluoroacetic acid

  • MDL number MFCD11007727
  • Cas Number : 475301-73-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart