LIF945291515 Life Chemicals Inc.

4-[(tert-butoxycarbonyl)amino]-2-(trifluoromethyl)benzoic acid

  • MDL number MFCD29037879
  • Cas Number : 1326229-55-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart