LIF945291520 Life Chemicals Inc.

methyl 7-amino-1,2,3,4-tetrahydroisoquinoline-2-carboxylate

  • MDL number MFCD23847531
  • Cas Number : 1448046-09-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart