LIF945291537 Life Chemicals Inc.

4-cyclopropyl-6-(piperazin-1-yl)pyrimidine

  • MDL number MFCD26382564
  • Cas Number : 1545121-65-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart