LIF945291538 Life Chemicals Inc.

6-(pyridin-2-yl)pyridazine-3-thiol

  • MDL number MFCD29999505
  • Cas Number : 1005036-25-8
  • Molecular Weight : 189.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart