LIF945291545 Life Chemicals Inc.

1-[3-(trifluoromethyl)phenyl]-1,3-diazinane-2,4,6-trione

  • MDL number MFCD01952047
  • Cas Number : 19136-42-6
  • Molecular Weight : 272.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart