LIF945291552 Life Chemicals Inc.

2-(5-chlorobenzo[d]isoxazol-3-yl)acetic acid

  • MDL number MFCD10008487
  • Cas Number : 59899-91-1
  • Molecular Weight : 211.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart