LIF945291557 Life Chemicals Inc.

4-chloro-6-(difluoromethyl)pyrimidine

  • MDL number MFCD18415625
  • Cas Number : 1261737-05-6
  • Molecular Weight : 164.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart