LIF945291569 Life Chemicals Inc.

3-fluoro-2,2-dimethylpropan-1-ol

  • MDL number MFCD29046498
  • Cas Number : 883444-33-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form oil
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart