LIF945291572 Life Chemicals Inc.

2-(1H-1,2,4-triazol-1-yl)nicotinaldehyde

  • MDL number MFCD23161394
  • Cas Number : 1374649-96-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart