LIF945291573 Life Chemicals Inc.

CIS-2-methoxycyclopropan-1-amine hydrochloride

  • MDL number MFCD19215476
  • Cas Number : 1046469-17-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart