LIF945291576 Life Chemicals Inc.

2-chloro-4-(1H-tetrazol-1-yl)pyridine

  • MDL number MFCD24387765
  • Cas Number : 1428233-50-4
  • Molecular Weight : 181.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart