LIF945291577 Life Chemicals Inc.

ethyl 5-amino-1-(3-chlorobenzyl)-1H-pyrazole-4-carboxylate

  • MDL number MFCD01202735
  • Cas Number : 1397219-57-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart