LIF945291581 Life Chemicals Inc.

2-(4-(pyridin-4-yl)-1H-pyrazol-1-yl)ethan-1-amine

  • MDL number MFCD30004129
  • Cas Number : 1864555-50-9
  • Molecular Weight : 188.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart