LIF945291584 Life Chemicals Inc.

1-(pyrrolidin-3-yl)piperidine-4-carboxamide dihydrochloride

  • MDL number MFCD29046800
  • Cas Number : 2095409-36-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart