LIF945291585 Life Chemicals Inc.

4-(pyrrolidin-3-yl)piperazin-2-one dihydrochloride

  • MDL number MFCD13561594
  • Cas Number : 1219957-44-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart