LIF945291588 Life Chemicals Inc.

2',2'-difluoro-8-azaspiro[bicyclo[3.2.1]octane-3,1'-cyclopropane] hydrochloride

  • MDL number MFCD30004251
  • Cas Number : 2095410-82-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart