LIF945291594 Life Chemicals Inc.

2-(3-(pyridin-4-yl)-1H-pyrazol-1-yl)acetonitrile

  • MDL number MFCD30005733
  • Cas Number : 2095410-75-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart