LIF945291595 Life Chemicals Inc.

methyl 3-(pyridin-3-yl)-1H-pyrazole-5-carboxylate

  • MDL number MFCD06166975
  • Cas Number : 898052-20-5
  • Molecular Weight : 203.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart