LIF945291597 Life Chemicals Inc.

methyl 3-(pyrazin-2-yl)-1H-pyrazole-5-carboxylate

  • MDL number MFCD30006137
  • Cas Number : 1873310-01-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart