LIF945291598 Life Chemicals Inc.

1-methyl-3-(pyrazin-2-yl)-1H-pyrazole-5-carboxamide

  • MDL number MFCD30006167
  • Cas Number : 2095410-85-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart