LIF945291599 Life Chemicals Inc.

3-(chloromethyl)-2-methyl-2,4,5,6-tetrahydrocyclopenta[c]pyrazole

  • MDL number MFCD30009988
  • Cas Number : 1251760-95-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form oil
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart