LIF945291600 Life Chemicals Inc.

3-(chloromethyl)-2-methyl-4,5,6,7-tetrahydro-2H-indazole

  • MDL number MFCD20628002
  • Cas Number : 741610-91-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart