LIF945291603 Life Chemicals Inc.

(1-methyl-2-pyrazolo[1,5-a]pyridin-3-ylethyl)amine

  • MDL number MFCD24387939
  • Cas Number : 1447963-42-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart