LIF945291604 Life Chemicals Inc.

5-acetamidopyrazolo[1,5-a]pyridine-3-carboxylic acid

  • MDL number MFCD26793391
  • Cas Number : 1706460-51-6
  • Molecular Weight : 219.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart