LIF945291609 Life Chemicals Inc.

4-(trifluoromethyl)pyrrolidine-3-carboxylic acid hydrochloride

  • MDL number MFCD11848054
  • Cas Number : 1260759-01-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart