LIF945291614 Life Chemicals Inc.

sodium 3-(4-chlorophenyl)-3-oxoprop-1-en-1-olate

  • MDL number MFCD04621494
  • Cas Number : 40160-35-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart