LIF945291618 Life Chemicals Inc.

1-bromoisoquinolin-4-amine

  • MDL number MFCD18448504
  • Cas Number : 55404-28-9
  • Molecular Weight : 223.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart