LIF945291632 Life Chemicals Inc.

ethyl 1-methyl-1H-1,2,3-triazole-4-carboxylate

  • MDL number MFCD08690701
  • Cas Number : 39039-48-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart